missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 6-Maleimidohexanoate, TRC

Product Code. 30412990
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412990 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412990 Supplier Toronto Research Chemicals Supplier No. TRCS68806510MG
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Succinimidyl 6-Maleimidohexanoate
CAS 55750-63-5
Molecular Formula C14H16N2O6
Quantity 10 mg
Synonym 6-Maleimidohexanoic Acid N-Hydroxysuccinimide Ester
InChI Formula InChI=1S/C14H16N2O6/c17-10-5-6-11(18)15(10)9-3-1-2-4-14(21)22-16-12(19)7-8-13(16)20/h5-6H,1-4,7-9H2
SMILES O=C(CCCCCN1C(=O)C=CC1=O)ON2C(=O)CCC2=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 6-(2,5-dioxopyrrol-1-yl)hexanoate
Molecular Weight (g/mol) 308.28
Formula Weight 308.1008
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.