missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 3-maleimidobenzoate, 95%

Product Code. p-7042123
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15404019

Brand: Thermo Scientific Alfa Aesar J61410.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A heterobifunctional coupling reagent useful for forming enzyme immunoconjugates | CAS: 58626-38-3 | C15H10N2O6 | 314.253 g/mol

N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

Solubility
Soluble in DMF or ethyl acetate.

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58626-38-3
Molecular Formula C15H10N2O6
Molecular Weight (g/mol) 314.253
MDL Number MFCD00005514
InChI Key LLXVXPPXELIDGQ-UHFFFAOYSA-N
Synonym 3-maleimidobenzoic acid n-hydroxysuccinimide ester, n-succinimidyl 3-maleimidobenzoate, 3-maleimidobenzoyl n-hydroxysuccinimide, 1-3-2,5-dioxopyrrolidinyl oxy carbonyl phenyl-1h-pyrrole-2,5-dione, m-maleimidobenzoyl-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl benzoate, 3-n-maleimidobenzoic acid n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl benzoate, ambotzpeg2090
PubChem CID 93861
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)benzoate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC(=CC=C2)N3C(=O)C=CC3=O

Specifications

Melting Point 175°C to 177°C
Color White
Quantity 250 mg
Beilstein 1505254
Sensitivity Light and moisture sensitive
Solubility Information Soluble in DMF or ethyl acetate.
Formula Weight 314.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Succinimidyl 3-maleimidobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.