missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate, 90+%
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.
Solubility
Soluble in dimethyl sulfoxide and acetonitrile.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate is an amine reactive reagent for azide incorporation. It contains a PEG spacer.
Solubility
Soluble in dimethyl sulfoxide and acetonitrile.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Color | Colorless to Yellow |
| Quantity | 25 mg |
| Solubility Information | Soluble in dimethyl sulfoxide and acetonitrile. |
| Formula Weight | 388.37 |
| Physical Form | Liquid |
| Chemical Name or Material | N-Succinimidyl 15-azido-4,7,10,13-tetraoxapentadecanoate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction