missing translation for 'onlineSavingsMsg'
Learn More

N-[(R)-alpha-(4-Chlorophenyl)benzyl]formamide, 98%, Thermo Scientific™

Catalog Number 15436646
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
474654-18-7
C14H12ClNO
245.71
HDGOSEHHSVUCBO-UHFFFAOYNA-N
n-r-phenyl 4-chlorophenyl methyl formamide, n-r-4-chlorophenyl phenylmethyl formamide, n-r-4-chlorophenyl phenyl methyl formamide
This item is not returnable. View return policy
474654-18-7
C14H12ClNO
245.71
HDGOSEHHSVUCBO-UHFFFAOYNA-N
n-r-phenyl 4-chlorophenyl methyl formamide, n-r-4-chlorophenyl phenylmethyl formamide, n-r-4-chlorophenyl phenyl methyl formamide

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 245.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-[(R)-alpha-(4-Chlorophenyl)benzyl]formamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'