missing translation for 'onlineSavingsMsg'
Learn More

N-Propylurea, 97%, Maybridge

Product Code. 10723663
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723663 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10723663 Supplier Thermo Scientific Maybridge Supplier No. TL00479EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 627-06-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C4H10N2O
Linear Formula CH3CH2CH2NHC(O)NH2
MDL Number MFCD00014793
Quantity 10 g
Synonym n-propylurea, 1-propylurea, urea, propyl, urea, n-propyl, propyl urea, unii-0pk51j4av7, n-propyl urea, n'-propylurea, amino-n-propylamide, acmc-1ato6
InChI Key ZQZJKHIIQFPZCS-UHFFFAOYSA-N
SMILES CCCNC(=O)N
IUPAC Name propylurea
Molecular Weight (g/mol) 102.137
PubChem CID 12303
Formula Weight 102.14
Percent Purity 97%
Chemical Name or Material N-Propylurea
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.