missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltriphenylphosphonium bromide, 99%, Thermo Scientific™

Product Code. 10625402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10625402 25 g Plastic bottle 25g
10397420 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10625402 Supplier Thermo Scientific Acros Supplier No. 157630250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15912-75-1
Molecular Formula C21H22P
Molecular Weight (g/mol) 305.38
MDL Number MFCD00011843
InChI Key MSXNDXIAUQJHNS-UHFFFAOYSA-N
Synonym propyl triphenyl phosphonium bromide, propyltriphenylphosphonium bromide, triphenylpropylphosphonium bromide at
PubChem CID 88526383
IUPAC Name triphenyl(propyl)phosphanium;hydrobromide
SMILES CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 235.0°C to 239.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula CH3(CH2)2P(C6H5)3Br
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform and polar solvents
Formula Weight 385.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material n-Propyltriphenylphosphonium bromide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • n-Propyltriphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.