missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltrimethoxysilane, 98+%

Product Code. 11420127
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420127 500 g 500g
11400127 25 g 25g
11410127 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420127 Supplier Thermo Scientific Alfa Aesar Supplier No. B21033.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1067-25-0 | C6H16O3Si | 164.28 g/mol

It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an important material in sol-gel production, provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst component for polyolefin production with Ziegler-Natta catalyst.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-25-0
Molecular Formula C6H16O3Si
Molecular Weight (g/mol) 164.28
MDL Number MFCD00043026
InChI Key HQYALQRYBUJWDH-UHFFFAOYSA-N
Synonym trimethoxy propyl silane, n-propyltrimethoxysilane, propyltrimethoxysilane, silane, trimethoxypropyl, unii-t72c7n9xo2, dynasylan ptmo, trimethoxysilylpropane, propyl-trimethoxysilane, silane,trimethoxypropyl, trimethoxy-n-propylsilane
PubChem CID 61254
IUPAC Name trimethoxy(propyl)silane
SMILES CCC[Si](OC)(OC)OC

Specifications

Boiling Point 142°C to 143°C
Quantity 500 g
UN Number UN1993
Beilstein 2070458
Flash Point 34°C (93°F)
Solubility Information Reacts with water.
Formula Weight 164.28
Percent Purity ≥98%
Refractive Index 1.39
Sensitivity Moisture sensitive
Density 0.935
Chemical Name or Material n-Propyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.