missing translation for 'onlineSavingsMsg'
Learn More

N-Propylethylenediamine, 99%

Product Code. 10651833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10651833 25 g Glass bottle 25g
10195850 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10651833 Supplier Thermo Scientific Acros Supplier No. 314750250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-39-7 | C5H14N2 | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-39-7
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008172
InChI Key CFNHVUGPXZUTRR-UHFFFAOYSA-N
Synonym n-propylethylenediamine, n-n-propyl ethylenediamine, 1,2-ethanediamine, n-propyl, ethylenediamine, n-propyl, 2-aminoethyl propyl amine, 1,2-ethanediamine, n1-propyl, 2-n-propylamino ethylamine, 2-n-propylaminoethylamine, propylethylenediamine, n-n-propylethylenediamine
PubChem CID 66073
IUPAC Name N'-propylethane-1,2-diamine
SMILES CCCNCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8200g/mL
Boiling Point 147°C to 150°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2CH2NHCH2CH2NH2
Refractive Index 1.4394 to 1.4414
Quantity 25 g
Specific Gravity 0.82
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Propylethylenediamine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.