missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl methacrylate, 95%, stab. with 200ppm 4-methoxyphenol

Product Code. p-7045123
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11367456

Brand: Thermo Scientific Alfa Aesar L09493.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2210-28-8 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2210-28-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00048690
InChI Key NHARPDSAXCBDDR-UHFFFAOYSA-N
Synonym propyl methacrylate, n-propyl methacrylate, methacrylic acid, propyl ester, propyl 2-methyl-2-propenoate, 2-propenoic acid, 2-methyl-, propyl ester, unii-95q6j7a6n3, ccris 4864, methacrylic acid propyl, propyl 2-methylacrylate, propyl 2-methylacrylate #
PubChem CID 16638
IUPAC Name propyl 2-methylprop-2-enoate
SMILES CCCOC(=O)C(C)=C

Specifications

Density 0.902
Boiling Point 140°C to 141°C
Flash Point 30°C (86°F)
Odor Pungent
Refractive Index 1.419
Quantity 5 g
UN Number UN3272
Beilstein 1744849
Formula Weight 128.17
Percent Purity 95%
Chemical Name or Material n-Propyl methacrylate, stabilized with 200ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.