missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl isocyanate, 99%, Thermo Scientific Chemicals

Product Code. 10455822
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-78-1
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00002045
InChI Key OQURWGJAWSLGQG-UHFFFAOYSA-N
Synonym propyl isocyanate, propane, 1-isocyanato, n-propyl isocyanate, 1-propyl isocyanate, propylisocyanate, isocyanic acid, propyl ester, m-propyl isocyanate, isocyanic acid propyl ester, 1-propylisocyanate, 1-isocyanato-propane
PubChem CID 61033
IUPAC Name 1-isocyanatopropane
SMILES CCCN=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color Yellow
Density 0.8948g/mL
Boiling Point 83.0°C to 84.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2NCO
Refractive Index 1.3940 to 1.3970
Quantity 25 g
Beilstein 04,I,366
Specific Gravity 0.8948
Solubility Information Solubility in water: reacts vigorously
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Propyl isocyanate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Propyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'