missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl isocyanate, 99%

Product Code. 10183660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183660 100 g Glass bottle 100g
10680463 5 g Glass bottle 5g
10455822 25 g Glass bottle 25g
10541381 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10183660 Supplier Thermo Scientific Acros Supplier No. 131601000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 110-78-1 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-78-1
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00002045
InChI Key OQURWGJAWSLGQG-UHFFFAOYSA-N
Synonym propyl isocyanate, propane, 1-isocyanato, n-propyl isocyanate, 1-propyl isocyanate, propylisocyanate, isocyanic acid, propyl ester, m-propyl isocyanate, isocyanic acid propyl ester, 1-propylisocyanate, 1-isocyanato-propane
PubChem CID 61033
IUPAC Name 1-isocyanatopropane
SMILES CCCN=C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color Yellow
Density 0.8948g/mL
Boiling Point 83.0°C to 84.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2NCO
Refractive Index 1.3940 to 1.3970
Quantity 100 g
Beilstein 04,I,366
Specific Gravity 0.8948
Solubility Information Solubility in water: reacts vigorously
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Propyl isocyanate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Propyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.