missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Ether, 99+%

Product Code. 10123580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10123580 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10123580 Supplier Thermo Scientific Acros Supplier No. 150040500
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-43-3 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-43-3
CAS Min % 99.0
CAS Max % 100.0
Melting Point -123°C
Color Colorless
Density 0.7300g/mL
Boiling Point 88°C to 90°C
Flash Point -28°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C6H14O
Linear Formula (CH3CH2CH2)2O
Refractive Index 1.3790 to 1.381
MDL Number MFCD00009376
Quantity 50 mL
Beilstein 01, 354
Merck Index 15, 3385
Specific Gravity 0.73
Synonym dipropyl ether, propyl ether, n-propyl ether, di-n-propyl ether, propane, 1,1'-oxybis, dipropyl oxide, 4-oxaheptane, ether, di-n-propyl, 1,1'-oxybispropane, unii-42q250hs4g
Solubility Information Solubility in water: in water: 3g/L (20°C). Other solubilities: soluble in alcohol,ether
InChI Key POLCUAVZOMRGSN-UHFFFAOYSA-N
SMILES CCCOCCC
IUPAC Name 1-propoxypropane
Molecular Weight (g/mol) 102.18
PubChem CID 8114
Formula Weight 102.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Propyl ether, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Propyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.