missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl bromoacetate, 97%

Product Code. 11378515
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378515 25 g 25g
11368515 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378515 Supplier Thermo Scientific Alfa Aesar Supplier No. L11951.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35223-80-4 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35223-80-4
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00075247
InChI Key ISYUCUGTDNJIHV-UHFFFAOYSA-N
Synonym propyl bromoacetate, n-propyl bromoacetate, acetic acid, bromo-, propyl ester, acmc-1ajeq, bromoacetic acid propyl ester, bromo-acetic acid propyl ester, acetic acid, 2-bromo-,propyl ester
PubChem CID 141981
IUPAC Name propyl 2-bromoacetate
SMILES CCCOC(=O)CBr

Specifications

Density 1.399
Boiling Point 175°C to 176°C
Flash Point 82°C (179°F)
Refractive Index 1.451
Quantity 25 g
UN Number UN2922
Beilstein 1745564
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material n-Propyl bromoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.