missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl acetate, 99%

Product Code. 11388246
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388246 500 mL 500mL
11398246 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388246 Supplier Thermo Scientific Alfa Aesar Supplier No. L15355.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-60-4 | C5H10O2 | 102.13 g/mol

n-propyl acetate is used as a solvent. It plays an important role in the printing inks industry are flexographic and special screen printing inks. It is widely used in fragrances and as a flavor additive due to its odor. It acts as a good solvent for cellulose nitrate, acrylates, alkyd resins, rosin, plasticizers, waxes, oils and fats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-propyl acetate is used as a solvent. It plays an important role in the printing inks industry are flexographic and special screen printing inks. It is widely used in fragrances and as a flavor additive due to its odor. It acts as a good solvent for cellulose nitrate, acrylates, alkyd resins, rosin, plasticizers, waxes, oils and fats.

Solubility
Miscible with alcohols, ketones, aldehydes, ethers, glycols and glycol ethers. Slightly soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009372
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
PubChem CID 7997
ChEBI CHEBI:40116
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

Melting Point -92°C
Density 0.887
Boiling Point 99°C to 102°C
Flash Point 14°C (57°F)
Odor Fruit-like
Linear Formula CH3CO2CH2CH2CH3
Refractive Index 1.384
Quantity 500 mL
UN Number UN1276
Beilstein 1740764
Merck Index 14,7841
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,glycols and glycol ethers. Slightly soluble in water.
Formula Weight 102.13
Percent Purity 99%
Chemical Name or Material n-Propyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Propyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: n-Propyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.