missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl acetate, 99%, pure

Product Code. 10215920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10215920 1 L Glass bottle 1 L
10449450 2.5 L Glass bottle 2.5 L
10194093 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215920 Supplier Thermo Scientific Acros Supplier No. 158290010
  • ONLINE EXCLUSIVE PRICE
    £47.00
    £45.60 / 1 L
    Save £1.40
    2% Off
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-60-4 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009372
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
PubChem CID 7997
ChEBI CHEBI:40116
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -95.0°C
Density 0.8800g/mL
Boiling Point 102.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2CH3
Refractive Index 1.3830 to 1.3850
Quantity 1 L
UN Number 1276
Beilstein 02, 129
Merck Index 15, 7954
Specific Gravity 0.88
Solubility Information Solubility in water: 2g/100mL (20°C). Other solubilities: miscible with alcohols,ketones,esters and,hydrocarbons
Viscosity 0.58 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Propyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Propyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.