missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl acetate, 99%, pure, Thermo Scientific Chemicals

Product Code. 10449450
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009372
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
PubChem CID 7997
ChEBI CHEBI:40116
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -95.0°C
Density 0.8800g/mL
Boiling Point 102.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2CH3
Refractive Index 1.3830 to 1.3850
Quantity 2.5 L
UN Number 1276
Beilstein 02, 129
Merck Index 15, 7954
Specific Gravity 0.88
Solubility Information Solubility in water: 2g/100mL (20°C). Other solubilities: miscible with alcohols,ketones,esters and,hydrocarbons
Viscosity 0.58 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Propyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Propyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'