missing translation for 'onlineSavingsMsg'
Learn More

N-(Propargyloxy)phthalimide, TRC

Product Code. 30425781
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30425781 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30425781

Brand: Toronto Research Chemicals TRCP83918310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(Propargyloxy)phthalimide
CAS 4616-63-1
Molecular Formula C11H7NO3
Quantity 10 mg
Synonym 2-prop-2-ynoxyisoindoline-1,3-dione
InChI Formula InChI=1S/C11H7NO3/c1-2-7-15-12-10(13)8-5-3-4-6-9(8)11(12)14/h1,3-6H,7H2
SMILES C#CCON1C(=O)C2=CC=CC=C2C1=O
IUPAC Name 2-prop-2-ynoxyisoindole-1,3-dione
Molecular Weight (g/mol) 201.18
Formula Weight 201.0426
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.