missing translation for 'onlineSavingsMsg'
Learn More

n-Propanol (Peroxide-Free/Sequencing), Fisher BioReagents

Product Code. 10326503
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10326503 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10326503 Supplier Fisher BioReagents Supplier No. BP1130500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
n-Propanol is used for extracting ethidium bromide from DNA samples.
TRUSTED_SUSTAINABILITY

Specifications

pH 7
Melting Point -127°C
CAS 71-23-8
Color Colorless
Acidity 0.02% max.
Density 0.800g/cm³
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.01% max.
Assay Percent Range ≥99.5 %
Molecular Formula C3H8O
Linear Formula CH3CH2CH2OH
Vapor Pressure 25mbar at 20°C
MDL Number MFCD00002941
Viscosity 2.2 mPa/s at 20°C
Merck Index 15, 7955
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
Boiling Point 97°C
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES CCCO
IUPAC Name propan-1-ol
Molecular Weight (g/mol) 60.10
PubChem CID 1031
ChEBI CHEBI:28831
Formula Weight 60.1
Percent Purity ≥99.5%
Grade Sequencing
Chemical Name or Material n-Propanol, Peroxide-free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.