missing translation for 'onlineSavingsMsg'
Learn More

N-[(Phenylmethoxy)carbonyl]-L-serine Methyl Ester, TRC

Product Code. 30410629
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410629 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410629 Supplier Toronto Research Chemicals Supplier No. TRCP33572525G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(Phenylmethoxy)carbonyl]-L-serine Methyl Ester
CAS 1676-81-9
Molecular Formula C12H15NO5
Quantity 25 g
Synonym N-Benzyl-N-carboxyserine Methyl Ester, N-Benzyl-N-carboxy-L-serine Methyl Ester, (2S)-2-[(Benzyloxycarbonyl)amino]-3-hydroxypropionic Acid Methyl Ester, (2S)-2-[(Benzyloxycarbonyl)amino]-3-hydroxypropionic Acid Methyl Ester, (Benzyloxycarbonyl)-L-serine Methyl Ester, Methyl (2S)-2-[(Benzyloxycarbonyl)amino]-3-hydroxypropanoate, Methyl (S)-2-((Benzyloxycarbonyl)amino)-3-hydroxypropanoate, Methyl N-(Benzyloxycarbonyl)-L-serinate, N-(Benzyloxycarbonyl)-L-serine Methyl Ester, N-(Benzyloxycarbonyl)serine Methyl Ester, N-Carbobenzoxy-L-serine Methyl Ester
InChI Formula InChI=1S/C12H15NO5/c1-17-11(15)10(7-14)13-12(16)18-8-9-5-3-2-4-6-9/h2-6,10,14H,7-8H2,1H3,(H,13,16)/t10-/m0/s1
SMILES COC(=O)[C@H](CO)NC(=O)OCc1ccccc1
IUPAC Name methyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
Molecular Weight (g/mol) 253.25
Formula Weight 253.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.