missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylmethanesulfonamide, 98%, Thermo Scientific™

Product Code. 10722222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722222 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10722222 Supplier Thermo Scientific Acros Supplier No. 417640025
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1197-22-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 93°C to 97°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H9NO2S
Linear Formula CH3SO2NHC6H5
MDL Number MFCD00043782
Quantity 2.5 g
Synonym methanesulfonamide, n-phenyl, methanesulfonanilide, methanesulfanilide, mesylaniline, n-mesylaniline, n-methylsulphonylaniline, n-phenyl-methanesulfonamide, methylsulfonyl phenylamine, mesylanilide, methylsulfonyl aniline
InChI Key LBTPIFQNEKOAIM-UHFFFAOYSA-N
SMILES CS(=O)(=O)NC1=CC=CC=C1
IUPAC Name N-phenylmethanesulfonamide
Molecular Weight (g/mol) 171.21
PubChem CID 70970
Formula Weight 171.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylmethanesulfonamide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.