missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylmaleimide, 97%, Thermo Scientific Chemicals

Catalog Number p-182750
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
941-69-5
C10H7NO2
173.17
MFCD00005502
HIDBROSJWZYGSZ-UHFFFAOYSA-N
n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
13662
1-phenylpyrrole-2,5-dione
C1=CC=C(C=C1)N2C(=O)C=CC2=O
This item is not returnable. View return policy
941-69-5
C10H7NO2
173.17
MFCD00005502
HIDBROSJWZYGSZ-UHFFFAOYSA-N
n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
13662
1-phenylpyrrole-2,5-dione
C1=CC=C(C=C1)N2C(=O)C=CC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 86°C to 90°C
Color Yellow
Boiling Point 162°C to 163°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 400
Fieser 01,845; 13,317
Merck Index 15, 7410
Formula Weight 173.17
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material N-Phenylmaleimide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N-Phenylmaleimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'