missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyliminodiacetic acid, 97%

Product Code. 11398165
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398165 25 g 25g
11388165 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398165 Supplier Thermo Scientific Alfa Aesar Supplier No. L08437.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1137-73-1 | C10H11NO4 | 209.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-73-1
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00043570
InChI Key GQBWTAGIANQVGB-UHFFFAOYSA-N
Synonym n-phenyliminodiacetic acid, glycine, n-carboxymethyl-n-phenyl, acetic acid,1-phenyliminodi, 2,2'-phenylazanediyl diacetic acid, 2,2'-phenylimino diacetic acid, n-carboxymethyl-n-phenylglycine, n,n-anilinediacetic acid, n-phenyliminodiacetic acid anhydride, nn-di carboxymethyl aniline, glycine,n-carboxymethyl-n-phenyl
PubChem CID 136915
IUPAC Name 2-[N-(carboxymethyl)anilino]acetic acid
SMILES C1=CC=C(C=C1)N(CC(=O)O)CC(=O)O

Specifications

Melting Point ∽140°C (decomposition)
Quantity 25 g
Beilstein 2808435
Formula Weight 209.2
Percent Purity 97%
Chemical Name or Material N-Phenyliminodiacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.