missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylglycine ethyl ester, 99%

Product Code. 11475946
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475946 50 g 50g
11465946 10 g 10g
11485946 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475946 Supplier Thermo Scientific Alfa Aesar Supplier No. B22282.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2216-92-4 | C10H13NO2 | 179.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-92-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00038315
InChI Key MLSGRWDEDYJNER-UHFFFAOYSA-N
Synonym n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u
PubChem CID 75190
IUPAC Name ethyl 2-anilinoacetate
SMILES CCOC(=O)CNC1=CC=CC=C1

Specifications

Melting Point 55°C to 58°C
Merck Index 14,7292
Quantity 50 g
Formula Weight 179.22
Percent Purity 99%
Chemical Name or Material N-Phenylglycine ethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.