missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylglycine ethyl ester, 99%, Thermo Scientific™

Product Code. 10127710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127710 50 g Glass bottle 50g
10531892 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10127710 Supplier Thermo Scientific Acros Supplier No. 417570500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2216-92-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00038315
InChI Key MLSGRWDEDYJNER-UHFFFAOYSA-N
Synonym n-phenylglycine ethyl ester, ethyl phenylglycinate, ethyl 2-phenylamino acetate, ethyl n-phenylglycinate, glycine, n-phenyl-, ethyl ester, unii-7xl9t8bk0u, ethyl anilinoacetate, n-phenylglycineethylester, n-phenylglycine ethyl, 7xl9t8bk0u
PubChem CID 75190
IUPAC Name ethyl 2-anilinoacetate
SMILES CCOC(=O)CNC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 g
Merck Index 15, 7403
Formula Weight 179.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylglycine ethyl ester, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.