missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylethanolamine, 98%

Product Code. 11309759
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309759 500 g 500g
11399749 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309759 Supplier Thermo Scientific Alfa Aesar Supplier No. B22131.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-98-5 | C8H11NO | 137.182 g/mol

N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5'-diphospho-glucuronosyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5′-diphospho-glucuronosyltransferase.

Solubility
Slightly miscible with water.

Notes
Incompatible with acids, acid anhydrides, acid chlorides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-98-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00002832
InChI Key MWGATWIBSKHFMR-UHFFFAOYSA-N
Synonym 2-phenylamino ethanol, n-phenylethanolamine, n-2-hydroxyethyl aniline, ethanol, 2-phenylamino, 2-phenylamino ethan-1-ol, ethanol, 2-anilino, beta-anilinoethanol, aniline, n-2-hydroxyethyl, n-2-hydroxyethyl phenylamine, emery 5700
PubChem CID 31235
IUPAC Name 2-anilinoethanol
SMILES C1=CC=C(C=C1)NCCO

Specifications

Melting Point -30°C
Density 1.087
pH 10.2
Boiling Point 284°C to 286°C
Flash Point 153°C (307°F)
Odor Amine-like
Refractive Index 1.579
Quantity 500 g
UN Number UN2810
Beilstein 774672
Solubility Information Slightly miscible with water.
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material N-Phenylethanolamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.