missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylanthranilic acid, 99%

Product Code. 11426907
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426907 500 g 500g
11406907 25 g 25g
11416907 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426907 Supplier Thermo Scientific Alfa Aesar Supplier No. A14340.36
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-40-7 | C13H11NO2 | 213.236 g/mol

DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.

Solubility
Soluble in ethanol, DMSO (50 mg/ml), hot alcohol, and methanol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-40-7
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00002421
InChI Key ZWJINEZUASEZBH-UHFFFAOYSA-N
Synonym n-phenylanthranilic acid, 2-phenylamino benzoic acid, fenamic acid, diphenylamine-2-carboxylic acid, phenylanthranilic acid, 2-carboxydiphenylamine, benzoic acid, 2-phenylamino, o-anilinobenzoic acid, n-phenyl-o-aminobenzoic acid, n-phenyl-2-aminobenzoic acid
PubChem CID 4386
ChEBI CHEBI:34756
IUPAC Name 2-anilinobenzoic acid
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 185°C to 188°C
Quantity 500 g
Beilstein 1456607
Merck Index 14,7273
Solubility Information Soluble in ethanol,DMSO (50mg/ml),hot alcohol,and methanol. Insoluble in water.
Formula Weight 213.24
Percent Purity 99%
Chemical Name or Material N-Phenylanthranilic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.