missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylacetamide (Acetanilide), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-Phenylacetamide (Acetanilide)
Specifications
Specifications
| Chemical Name or Material | N-Phenylacetamide |
| CAS | 103-84-4 |
| Analyte or Component Names | N-Phenylacetamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H9 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-Phenylacetamide, Paracetamol Imp. D (EP), Acetanilide, Acetamidobenzene, Acetanil, Acetoanilide, Acetylaniline, Antifebrin, Benzenamine, N-acetyl-, N-Acetylaminobenzene, N-Acetylaniline, NSC 203231, NSC 7636, Phenalgene, Phenalgin, Acetaminophen USP Related Compound D, Acetaminophen USP RC D, Acetaminophen Related Compound D, Paracetamol Impurity D |
| InChI Formula | InChI=1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10) |
| SMILES | CC(=O)Nc1ccccc1 |
| IUPAC Name | N-phenylacetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction