missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-p-phenylenediamine, 98%

Product Code. 10400691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10400691 250 g Glass bottle 250g
10696042 100 g Glass bottle 100g
10490681 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10400691 Supplier Thermo Scientific Acros Supplier No. 221002500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-54-2 | C12H12N2 | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-54-2
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00007850
InChI Key ATGUVEKSASEFFO-UHFFFAOYSA-N
Synonym 4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine
PubChem CID 7564
ChEBI CHEBI:59038
IUPAC Name 4-N-phenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.0°C to 75.0°C
Boiling Point 354.0°C
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NHC6H4NH2
Quantity 250 g
Beilstein 13, 76
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 184.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Phenyl-p-phenylenediamine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Phenyl-p-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.