Learn More
N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals
£25.30 - £201.00
Chemical Identifiers
CAS | 101-54-2 |
---|---|
Molecular Formula | C12H12N2 |
Molecular Weight (g/mol) | 184.24 |
MDL Number | MFCD00007850 |
InChI Key | ATGUVEKSASEFFO-UHFFFAOYSA-N |
Synonym | 4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine |
PubChem CID | 7564 |
ChEBI | CHEBI:59038 |
IUPAC Name | 4-N-phenylbenzene-1,4-diamine |
SMILES | C1=CC=C(C=C1)NC2=CC=C(C=C2)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10696042
|
Thermo Scientific Acros
221001000 |
100 g | Glass bottle |
£25.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10400691
|
Thermo Scientific Acros
221002500 |
250 g | Glass bottle |
£56.60
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10490681
|
Thermo Scientific Acros
221000010 |
1 kg | Glass bottle |
£201.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
101-54-2 | |
184.24 | |
ATGUVEKSASEFFO-UHFFFAOYSA-N | |
7564 | |
4-N-phenylbenzene-1,4-diamine |
C12H12N2 | |
MFCD00007850 | |
4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine | |
CHEBI:59038 | |
C1=CC=C(C=C1)NC2=CC=C(C=C2)N |
Specifications
101-54-2 | |
100.0 | |
354.0°C | |
Authentic | |
C12H12N2 | |
MFCD00007850 | |
4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine | |
ATGUVEKSASEFFO-UHFFFAOYSA-N | |
4-N-phenylbenzene-1,4-diamine | |
7564 | |
184.24 | |
Liquid |
97.5 | |
69.0°C to 75.0°C | |
>250°C | |
97.5% min. (GC) | |
C6H5NHC6H4NH2 | |
13, 76 | |
Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in alcohol and acetone | |
C1=CC=C(C=C1)NC2=CC=C(C=C2)N | |
184.24 | |
CHEBI:59038 | |
98% | |
N-Phenyl-p-phenylenediamine, 98% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause an allergic skin reaction.
Harmful if swallowed.
Very toxic to aquatic life
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-951-9
missing translation for 'rtecsNumber' : ST3150000
missing translation for 'tsca' : TSCA
- N-Phenyl-p-phenylenediamine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only