missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-2-naphthylamine, TRC

Product Code. 30424164
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424164 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424164 Supplier Toronto Research Chemicals Supplier No. TRCP3361455G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-88-6
Molecular Formula C16 H13 N
Molecular Weight (g/mol) 219.28
Synonym 2-(Phenylamino)naphthalene, 2-Anilinonaphthalene, 2-Naphthylphenylamine, AK 1, AK 1 (stabilizer), Aceto PBN, AgeRite Powder, Antioxidant 116, Antioxidant D, Antioxidant PBN, N-(2-Naphthyl)-N-phenylamine, N-(2-Naphthyl)aniline, N-Phenyl-2-naphthalenamine, N-Phenyl-2-naphthylamine, N-Phenyl-N-(2-naphthyl)amine, N-Phenyl-β-naphthylamine, N-β-Naphthyl-N-phenylamine, NSC 37151, Naftam 2, Neosone D, Neozon D, Neozone, Neozone D, Nilox PBNA, Noclizer D, Nocrac D, Nonox D, Nonox DN, PBNA, PNA, Phenyl-2-naphthylamine, Phenyl-β-naphthylamine, Stabilizator AR, Stabilizer AR, Vulkanox PBN, β-Naphthylphenylamine
IUPAC Name N-phenylnaphthalen-2-amine
SMILES N(c1ccccc1)c2ccc3ccccc3c2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H13N/c1-2-8-15(9-3-1)17-16-11-10-13-6-4-5-7-14(13)12-16/h1-12,17H
Formula Weight 219.1048
Chemical Name or Material N-Phenyl-2-naphthylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.