missing translation for 'onlineSavingsMsg'
Learn More

N-Phenethylbenzamide, TRC

Product Code. 30409133
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409133 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409133 Supplier Toronto Research Chemicals Supplier No. TRCP2960505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3278-14-6
Molecular Formula C15 H15 N O
Molecular Weight (g/mol) 225.29
Synonym Benzamide, N-(2-phenylethyl)-, Benzamide, N-phenethyl- (6CI,7CI,8CI), N-(2-Phenylethyl)benzamide, N-(2-Phenylethyl)benzamide, N-Benzoyl-2-phenylethylamine, N-Benzoyl-β-phenethylamine, N-Benzoylphenethylamine, N-Phenethylbenzamide, NSC 16618, NSC 43723
IUPAC Name N-(2-phenylethyl)benzamide
SMILES O=C(NCCc1ccccc1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H15NO/c17-15(14-9-5-2-6-10-14)16-12-11-13-7-3-1-4-8-13/h1-10H,11-12H2,(H,16,17)
Formula Weight 225.1154
Chemical Name or Material N-(2-Phenylethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.