missing translation for 'onlineSavingsMsg'
Learn More

N-Phenacylpyridinium bromide, 97%, Thermo Scientific™

Product Code. 15770879 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15770879 10 g 10g
15780879 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15770879 Supplier Thermo Scientific Acros Supplier No. 450120100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16883-69-5
Molecular Formula C13H12BrNO
Molecular Weight (g/mol) 278.15
InChI Key PXSUMUYPXZEXDT-UHFFFAOYSA-M
Synonym 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide
PubChem CID 2798954
IUPAC Name 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide
SMILES C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-]

Specifications

Melting Point 193.0°C to 195.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Quantity 10 g
Solubility Information Solubility in water: soluble.
Formula Weight 278.15
Percent Purity 97%
Chemical Name or Material N-Phenacylpyridinium bromide
compliance-icons
Product Identifier
  • N-Phenacylpyridinium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1-(2-Oxo-2-phenylethyl)pyridinium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.