missing translation for 'onlineSavingsMsg'
Learn More

n-Pentylbenzene, 96%

Product Code. 11492368
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492368 25 g 25g
11408770 5 g 5g
11402378 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492368 Supplier Thermo Scientific Alfa Aesar Supplier No. A15734.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 538-68-1 | C11H16 | 148.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-68-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00009502
InChI Key PWATWSYOIIXYMA-UHFFFAOYSA-N
Synonym 1-phenylpentane, n-pentylbenzene, amylbenzene, n-amylbenzene, benzene, pentyl, 1-pentylbenzene, 1-phenyl-n-pentane, pentane, 1-phenyl, pentyl-benzene, pentane, phenyl
PubChem CID 10864
IUPAC Name pentylbenzene
SMILES CCCCCC1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.863
Boiling Point 204°C to 205°C
Flash Point 65°C (149°F)
Refractive Index 1.488
Quantity 25 g
UN Number UN3082
Beilstein 1904359
Merck Index 14,600
Formula Weight 148.25
Percent Purity 96%
Chemical Name or Material n-Pentylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.