missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentane, Spectrophotometric Grade, 99+% min., Thermo Scientific Chemicals
35.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Description
n-Pentane is used in spectrophotometry and environmental testing. It is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 130°C |
Color | Colorless |
Appearance | Colorless liquid |
Density | 0.626 |
Boiling Point | 36°C |
Quantity | 1 L |
Flash Point | -49°C (-57°F) |
Physical Form | Liquid |
Refractive Index | 1.358 |
Assay Percent Range | >99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'