missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, Spectrophotometric Grade, 99+% min.

Product Code. 11328387
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Unit Size:
1 L
16 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328387 4 L 4 L
11338387 1 L 1 L
11308387 4 x 1 L 4 L
11318387 4 x 4 L 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11328387 Supplier Thermo Scientific Alfa Aesar Supplier No. 032449.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 109-66-0 | C5H12 | 72.15 g/mol

n-Pentane is used in spectrophotometry and environmental testing. It is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point °C to 130°C
Color Colorless
Appearance Colorless liquid
Density 0.626
Quantity 4 L
Flash Point -49°C (-57°F)
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range >99%
Vapor Pressure 554 hPa (416mm Hg) at 20°C
Boiling Point 36°C
Solubility Information Miscible with water.
Formula Weight 72.15
Grade Spectrophotometry
Chemical Name or Material n-Pentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.