missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, HPLC Grade, 99% min

Product Code. 11368147
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368147 4 x 1 L 4 L
11348147 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11368147 Supplier Thermo Scientific Alfa Aesar Supplier No. 022907.M1
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Maximum level of impurities: Evaporation residue 3ppm, H2O 0.01% | CAS: 109-66-0 | C5H12 | 72.15 g/mol

n-Pentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point °C to 130°C
Color Colorless
Appearance Colorless liquid
Density 0.626
Quantity 4 x 1 L
Flash Point -49°C (-57°F)
Ignition Point 260°C (500°F)
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range 99% min.
Vapor Pressure 554 hPa (416mm Hg) at 20°C
Boiling Point 36°C
Solubility Information Miscible with water.
Formula Weight 72.15
Grade HPLC
Chemical Name or Material n-Pentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.