missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Pentane, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
2.5 L
25 L
5 L
500mL
Description
N-Pentane, Extra Pure, SLR
Specifications
Specifications
| Packaging | Metal can |
| Vapor Pressure | 573mbar at 20°C |
| Viscosity | 0.25 mPaS at 20°C |
| Vapor Density | 2.5 |
| Physical Form | Liquid |
| Melting Point | -130°C |
| Boiling Point | 36°C |
| Quantity | 25 L |
| Formula Weight | 72.15g/mol |
Safety and Handling
Product Identifier
- n-Pentane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction