missing translation for 'onlineSavingsMsg'
Learn More

N-Pentane, Certified AR for Analysis, Fisher Chemical™

Product Code. 10558250 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Unit Size:
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10558250 2.5 L Amber glass bottle 2.5 L
10366000 1 L Amber glass bottle 1 L
11458433 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10558250 Supplier Fisher Chemical Supplier No. P/1021/17
  • ONLINE EXCLUSIVE PRICE
    £386.00
    £226.00 / 2.5 L
    Save £160.00
    41% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

C5H12, CAS Number-109-66-0, 2.5L, 36 deg.C, CHEBI:37830, Undesignated, 72.15g/mol, OFBQJSOFQDEBGM-UHFFFAOYSA-N, pentane, -130 deg.C, 9498, 72.151, Amber glass bottle, Liquid, 8003, CCCCC, 573mbar at 20 deg.C, 0.25 mPaS at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Packaging Amber glass bottle
Vapor Pressure 573mbar at 20°C
Viscosity 0.25 mPaS at 20°C
Vapor Density 2.5
Physical Form Liquid
Melting Point -130°C
Boiling Point 36°C
Quantity 2.5 L
Formula Weight 72.15g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Pentane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.