missing translation for 'onlineSavingsMsg'
Learn More

n-Pentadecane, 99%

Product Code. 11484113
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
Unit Size:
250g
25g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484113 50 g 50g
11458720 25 g 25g
11494113 250 g 250g
11468720 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11484113 Supplier Thermo Scientific Alfa Aesar Supplier No. A10336.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-62-9 | C15H32 | 212.42 g/mol

n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-62-9
Molecular Formula C15H32
Molecular Weight (g/mol) 212.42
MDL Number MFCD00008990
InChI Key YCOZIPAWZNQLMR-UHFFFAOYSA-N
Synonym n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n
PubChem CID 12391
ChEBI CHEBI:28897
IUPAC Name pentadecane
SMILES CCCCCCCCCCCCCCC

Specifications

Melting Point 9°C to 10°C
Density 0.769
Boiling Point 269°C to 270°C
Flash Point 132°C (269°F)
Refractive Index 1.432
Quantity 50 g
Beilstein 1698194
Solubility Information Immiscible with water.
Formula Weight 212.42
Percent Purity 99%
Chemical Name or Material n-Pentadecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Pentadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Pentadecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.