missing translation for 'onlineSavingsMsg'
Learn More

N-PAC Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £366.00

Specifications

Antigen N-PAC
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18606881
View Documents
Novus Biologicals
NBP2-93036-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18615810
View Documents
Novus Biologicals
NBP2-93036-0.1ml
0.1 mL
£366.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-PAC Polyclonal antibody specifically detects N-PAC in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

N-PAC
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.3), 50% glycerol
84656
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
BM045, Cytokine-like nuclear factor N-PAC, Glyoxylate reductase 1 homolog, glyoxylate reductase 1 homolog (Arabidopsis), HIBDLnuclear protein 60 kDa, NP603-hydroxyisobutyrate dehydrogenase-like protein, N-PAC, nuclear protein 60kDa, Nuclear protein NP60, Nuclear protein of 60 kDa, putative oxidoreductase GLYR1
Recombinant fusion protein containing a sequence corresponding to amino acids 70-270 of human GLYR1 (NP_115958.2). MIKINKGKRFQQAVDAVEEFLRRAKGKDQTSSHNSSDDKNRRNSSEERSRPNSGDEKRKLSLSEGKVKKNMGEGKKRVSSGSSERGSKSPLKRAQEQSPRKRGRPPKDEKDLTIPESSTVKGMMAGPMAAFKWQPTASEPVKDADPHFHHFLLSQTEKPAVCYQAITKKLKICEEETGSTSIQAADSTAVNGSITPTDKKI
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.