missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)-L-phenylalanine chloromethyl ketone, Thermo Scientific™

Product Code. 15410147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410147 5 g 5g
15400147 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410147 Supplier Thermo Scientific Alfa Aesar Supplier No. J60639.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enzyme inhibitor that inactivates chymotrypsin but not trypsin

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-71-1
Molecular Formula C17H18ClNO3S
Molecular Weight (g/mol) 351.85
MDL Number MFCD00000935
InChI Key MQUQNUAYKLCRME-INIZCTEOSA-N
Synonym tpck, tos-phe-ch2cl, tosylphenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanyl chloromethyl ketone, n-tosyl-l-phenylalanine chloromethyl ketone, n-p-tosyl-l-phenylalanine chloromethyl ketone, l-1-tosylamido-2-phenylethyl chloromethyl ketone, l-n-alpha-chloroacetyl phenethyl-p-toluenesulfonamide, tos-phe-chloromethylketone, chembl60718
PubChem CID 439647
ChEBI CHEBI:9642
SMILES CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CC1=CC=CC=C1)C(=O)CCl

Specifications

Color White
Beilstein 2895215
Solubility Information Soluble in DMSO at 10mg/ml
IUPAC Name N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]-4-methylbenzene-1-sulfonamide
Quantity 5 g
Formula Weight 351.85
Physical Form Powder
Chemical Name or Material N-(p-Toluenesulfonyl)-L-phenylalanine chloromethyl ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.