missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Toluenesulfonyl)glycine, 97%

Product Code. 15460118
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460118 25 g 25g
15450118 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15460118 Supplier Thermo Scientific Alfa Aesar Supplier No. H53371.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1080-44-0 | C9H11NO4S | 229.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1080-44-0
Molecular Formula C9H11NO4S
Molecular Weight (g/mol) 229.25
MDL Number MFCD00045898
InChI Key VDKFCCZUCXYILI-UHFFFAOYSA-N
Synonym n-p-tosylglycine, tos-gly-oh, n-tosylglycine, 2-4-methylphenylsulfonamido acetic acid, n-4-methylphenyl sulfonyl glycine, n-p-toluenesulfonyl glycine, n-4-tosylglycine, glycine, n-4-methylphenyl sulfonyl, n-toluene-4-sulfonyl-glycine, toluene-4-sulfonylamino-acetic acid
PubChem CID 70653
IUPAC Name 2-[(4-methylphenyl)sulfonylamino]acetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCC(=O)O

Specifications

Melting Point 146°C to 149°C
Quantity 25 g
Formula Weight 229.25
Percent Purity 97%
Chemical Name or Material N-(p-Toluenesulfonyl)glycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.