missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Hydroxyphenethyl)-N-(3-hydroxy-4-methoxy)benzylamine, TRC

Product Code. 30397738
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397738 250 mg 250mg
30397777 1 g 1g
30401070 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397738 Supplier Toronto Research Chemicals Supplier No. TRCH948930250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4579-60-6
Molecular Formula C16 H19 N O3
Molecular Weight (g/mol) 273.33
IUPAC Name 5-[[2-(4-hydroxyphenyl)ethylamino]methyl]-2-methoxyphenol
SMILES COc1ccc(CNCCc2ccc(O)cc2)cc1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H19NO3/c1-20-16-7-4-13(10-15(16)19)11-17-9-8-12-2-5-14(18)6-3-12/h2-7,10,17-19H,8-9,11H2,1H3
Formula Weight 273.1365
Chemical Name or Material N-(p-Hydroxyphenethyl)-N-(3-hydroxy-4-methoxy)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.