missing translation for 'onlineSavingsMsg'
Learn More

N-(p-amylcinnamoyl) Anthranilic Acid, TRC

Product Code. 30341894
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30341894

Brand: Toronto Research Chemicals TRCA6365251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(p-amylcinnamoyl) Anthranilic Acid
CAS 110683-10-8
Molecular Formula C21H23NO3
Quantity 1 g
Synonym 2-[[1-Oxo-3-(4-pentylphenyl)-2-propen-1-yl]amino]-benzoic Acid, 2-[[1-oxo-3-(4-pentylphenyl)-2-propenyl]amino]-benzoic Acid, N-(4-Pentylcinnamoyl)anthranilic Acid
InChI Formula InChI=1S/C21H23NO3/c1-2-3-4-7-16-10-12-17(13-11-16)14-15-20(23)22-19-9-6-5-8-18(19)21(24)25/h5-6,8-15H,2-4,7H2,1H3,(H,22,23)(H,24,25)/b15-14+
SMILES CCCCCc1ccc(\C=C\C(=O)Nc2ccccc2C(=O)O)cc1
IUPAC Name 2-[[(E)-3-(4-pentylphenyl)prop-2-enoyl]amino]benzoic acid
Molecular Weight (g/mol) 337.41
Formula Weight 337.1678
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.