missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(p-amylcinnamoyl) Anthranilic Acid, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | N-(p-amylcinnamoyl) Anthranilic Acid |
| CAS | 110683-10-8 |
| Molecular Formula | C21H23NO3 |
| Quantity | 25 mg |
| Synonym | 2-[[1-Oxo-3-(4-pentylphenyl)-2-propen-1-yl]amino]-benzoic Acid, 2-[[1-oxo-3-(4-pentylphenyl)-2-propenyl]amino]-benzoic Acid, N-(4-Pentylcinnamoyl)anthranilic Acid |
| InChI Formula | InChI=1S/C21H23NO3/c1-2-3-4-7-16-10-12-17(13-11-16)14-15-20(23)22-19-9-6-5-8-18(19)21(24)25/h5-6,8-15H,2-4,7H2,1H3,(H,22,23)(H,24,25)/b15-14+ |
| SMILES | CCCCCc1ccc(\C=C\C(=O)Nc2ccccc2C(=O)O)cc1 |
| IUPAC Name | 2-[[(E)-3-(4-pentylphenyl)prop-2-enoyl]amino]benzoic acid |
| Molecular Weight (g/mol) | 337.41 |
| Formula Weight | 337.1678 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction