missing translation for 'onlineSavingsMsg'
Learn More

N-(p-amylcinnamoyl) Anthranilic Acid, MedChemExpress

Product Code. p-200098848
Encompass_Preferred
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16473276

Brand: Medchem Express HY118628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is a broad spectrum Phospholipase A2 (PLA2) inhibitor and TRP channel blocker. N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is also an effective reversible inhibitor of calcium-activated chloride channels, has potential to treat arrhythmia.

N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is a broad spectrum Phospholipase A2 (PLA2) inhibitor and TRP channel blocker. N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is also an effective reversible inhibitor of calcium-activated chloride channels, has potential to treat arrhythmia.

Specifications

Chemical Name or Material N-(p-amylcinnamoyl) Anthranilic Acid
Physical Form Solid
CAS 110683-10-8
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C21H23NO3
For Use With (Application) COVID-19-immunoregulation
Synonym ACA
Solubility Information DMSO : ≥ 125 mg/mL (370.47 mM)
SMILES CCCCCC1=CC=C(C=C1)/C=C/C(NC2=CC=CC=C2C(O)=O)=O
Molecular Weight (g/mol) 337.41
Formula Weight 337.41
Percent Purity 96.94%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.