missing translation for 'onlineSavingsMsg'
Learn More

N-(p-amylcinnamoyl) Anthranilic Acid, MedChemExpress

Product Code. 16473276
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473276 25 mg 25mg
16421147 50 mg 50mg
16405676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473276 Supplier Medchem Express Supplier No. HY118628
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is a broad spectrum Phospholipase A2 (PLA2) inhibitor and TRP channel blocker. N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is also an effective reversible inhibitor of calcium-activated chloride channels, has potential to treat arrhythmia.

N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is a broad spectrum Phospholipase A2 (PLA2) inhibitor and TRP channel blocker. N-(p-amylcinnamoyl) Anthranilic Acid (ACA) is also an effective reversible inhibitor of calcium-activated chloride channels, has potential to treat arrhythmia.

Specifications

Chemical Name or Material N-(p-amylcinnamoyl) Anthranilic Acid
Physical Form Solid
CAS 110683-10-8
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C21H23NO3
For Use With (Application) COVID-19-immunoregulation
Synonym ACA
Solubility Information DMSO : ≥ 125 mg/mL (370.47 mM)
SMILES CCCCCC1=CC=C(C=C1)/C=C/C(NC2=CC=CC=C2C(O)=O)=O
Molecular Weight (g/mol) 337.41
Formula Weight 337.41
Percent Purity 96.94%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.