missing translation for 'onlineSavingsMsg'
Learn More

N-Oleoylethanolamide, TRC

Product Code. 30135996
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135996 1 g 1g
30138361 50 mg 50mg
17962625 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135996 Supplier Toronto Research Chemicals Supplier No. TRCO5272801G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-58-0
Molecular Formula C20 H39 N O2
Molecular Weight (g/mol) 325.53
Synonym N-(2-Hydroxyethyl)-Oleamide, 9-Octadecenamide, N-(2-hydroxyethyl)-, (Z)-, Oleamide, N-(2-hydroxyethyl)- (6CI,7CI,8CI), (9Z)-N-(2-Hydroxyethyl)-9-octadecenamide, AM 3101, N-(2-Hydroxyethyl)-9-Z-octadecenamide, N-(2-Hydroxyethyl)oleamide, N-Oleoyl-2-aminoethanol, N-Oleoylethanolamide, N-Oleoylethanolamine, Oleamide MEA, Oleic acid ethanolamide, Oleic acid monoethanolamide, Oleoylethanolamide
IUPAC Name (Z)-N-(2-hydroxyethyl)octadec-9-enamide
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)NCCO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)21-18-19-22/h9-10,22H,2-8,11-19H2,1H3,(H,21,23)/b10-9-
Formula Weight 325.2981
Chemical Name or Material Oleoyl Ethanolamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.