missing translation for 'onlineSavingsMsg'
Learn More

n-Octylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15445365
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445365 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15445365 Supplier Thermo Scientific Alfa Aesar Supplier No. H58444.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 131379-13-0
Molecular Formula C8H17BrZn
Linear Formula CH3(CH2)7ZnBr
MDL Number MFCD07698727
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym n-octylzinc bromide, bromozinc 1+ ,octane, bromo octyl zinc, n-octylzinc bromide 0.5 m in tetrahydrofuran, n-octylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key KIWWBFIVWRIFIX-UHFFFAOYSA-M
SMILES CCCCCCC[CH2-].[Zn+]Br
IUPAC Name bromozinc(1+);octane
Molecular Weight (g/mol) 258.508
PubChem CID 14921096
Formula Weight 258.51
Chemical Name or Material n-Octylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.