missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltriethoxysilane, 95%

Product Code. 11301577
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301577 50 g 50g
11311577 250 g 250g
11321577 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11301577 Supplier Thermo Scientific Alfa Aesar Supplier No. L04407.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2943-75-1 | C14H32O3Si | 276.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2943-75-1
Molecular Formula C14H32O3Si
Molecular Weight (g/mol) 276.49
MDL Number MFCD00039883
InChI Key MSRJTTSHWYDFIU-UHFFFAOYSA-N
Synonym n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
PubChem CID 76262
IUPAC Name triethoxy(octyl)silane
SMILES CCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 265°C
Melting Point <-40°C
Quantity 50 g
Beilstein 2325287
Flash Point 109°C (228°F)
Formula Weight 276.49
Percent Purity 95%
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.878
Chemical Name or Material n-Octyltriethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.